Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081437.1 | NP_001074906.1 | 14 - 15 | 3061 | 52 | |
NM_007992.2 | NP_032018.2 | 15 - 16 | 3202 | 52 | ||
GenBank mRNA | AK087922.1 | - | 3 - 4 | 447 | 52 | |
AK159361.1 | - | 14 - 15 | 3373 | 52 | ||
AK161522.1 | - | 14 - 15 | 3004 | 52 | ||
AK168688.1 | - | 14 - 15 | 2999 | 52 | ||
BC005443.1 | - | 14 - 15 | 3041 | 3490759 | 52 | |
X75285.1 | - | 15 - 16 | 3102 | 52 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |