Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290424.1 | NP_001277353.1 | 6 - 7 | 792 | 86 | |
NM_008031.3 | NP_032057.2 | 6 - 7 | 792 | 86 | ||
GenBank mRNA | AF461114.1 | - | 6 - 7 | 516 | 86 | |
AK046605.1 | - | 6 - 7 | 752 | 86 | ||
AK053211.1 | - | 3 - 4 | 534 | 86 | ||
AK053701.1 | - | 6 - 7 | 752 | 86 | ||
AK053829.1 | - | 6 - 7 | 794 | 86 | ||
AK080948.1 | - | 6 - 7 | 752 | 86 | ||
AK140343.1 | - | 6 - 7 | 754 | 86 | ||
BC079671.1 | - | 6 - 7 | 778 | 30532682 | 86 | |
L23971.1 | - | 6 - 7 | 639 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |