Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008065.2 | NP_032091.2 | 6 - 7 | 1196 | 85 | |
GenBank mRNA | AK139916.1 | - | 6 - 7 | 876 | 85 | |
AK145446.1 | - | 6 - 7 | 876 | 85 | ||
AK145838.1 | - | 6 - 7 | 941 | 85 | ||
AK148087.1 | - | 6 - 7 | 967 | 85 | ||
AK150455.1 | - | 6 - 7 | 876 | 85 | ||
BC013562.1 | - | 4 - 5 | 591 | 3493987 | 85 | |
BC052448.1 | - | 6 - 7 | 1196 | 6408997 | 85 | |
M74515.1 | - | 6 - 7 | 1203 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |