Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008550.2 | NP_032576.2 | 10 - 11 | 1651 | 75 | |
GenBank mRNA | AB006458.1 | - | 10 - 11 | 1629 | 75 | |
AK004900.1 | - | 10 - 11 | 1605 | 75 | ||
AK040388.1 | - | 10 - 11 | 1612 | 75 | ||
AK075649.1 | - | 2 - 3 | 316 | 75 | ||
AK165376.1 | - | 10 - 11 | 1644 | 75 | ||
BC066211.1 | - | 10 - 11 | 1602 | 30101816 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |