Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008550.2 | NP_032576.2 | 14 - 15 | 2471 | 56 | |
GenBank mRNA | AB006458.1 | - | 14 - 15 | 2449 | 56 | |
AK004900.1 | - | 14 - 15 | 2425 | 56 | ||
AK040388.1 | - | 14 - 15 | 2432 | 56 | ||
AK075649.1 | - | 6 - 7 | 1138 | 56 | ||
AK079969.1 | - | 8 - 9 | 1019 | 56 | ||
AK165376.1 | - | 14 - 15 | 2464 | 56 | ||
BC066211.1 | - | 14 - 15 | 2422 | 30101816 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |