Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Men1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168488.1 | NP_001161960.1 | 5 - 6 | 928 | 75 | |
NM_001168489.1 | NP_001161961.1 | 5 - 6 | 906 | 75 | ||
NM_001168490.1 | NP_001161962.1 | 4 - 5 | 1242 | 75 | ||
NM_008583.2 | NP_032609.1 | 4 - 5 | 1242 | 75 | ||
GenBank mRNA | AB023401.1 | - | 4 - 5 | 869 | 75 | |
AF016398.1 | - | 5 - 6 | 881 | 75 | ||
AF072755.1 | - | 5 - 6 | 865 | 75 | ||
AF109389.1 | - | 4 - 5 | 1241 | 75 | ||
AF130368.1 | - | 4 - 5 | 1224 | 75 | ||
AK133984.1 | - | 5 - 6 | 865 | 75 | ||
AK154371.1 | - | 5 - 6 | 887 | 75 | ||
AK164700.1 | - | 5 - 6 | 863 | 75 | ||
AK170713.1 | - | 4 - 5 | 1222 | 75 | ||
BC006610.1 | - | 5 - 6 | 861 | 75 | ||
BC036287.1 | - | 4 - 5 | 1170 | 4189611 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |