Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mme | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289462.1 | NP_001276391.1 | 9 - 10 | 1312 | 100 | |
NM_001289463.1 | NP_001276392.1 | 9 - 10 | 1096 | 100 | ||
NM_008604.4 | NP_032630.2 | 9 - 10 | 946 | 100 | ||
GenBank mRNA | AK031446.1 | - | 9 - 10 | 947 | 100 | |
AK033824.1 | - | 9 - 10 | 1007 | 100 | ||
AK040730.1 | - | 9 - 10 | 984 | 101 | ||
AK052088.1 | - | 8 - 9 | 833 | 100 | ||
AK076908.1 | - | 9 - 10 | 1160 | 100 | ||
BC034092.1 | - | 9 - 10 | 968 | 4483558 | 100 | |
BC066840.1 | - | 8 - 9 | 855 | 100 | ||
M81591.1 | - | 8 - 9 | 858 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |