Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mme | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289462.1 | NP_001276391.1 | 18 - 19 | 2235 | 86 | |
NM_001289463.1 | NP_001276392.1 | 18 - 19 | 2019 | 86 | ||
NM_008604.4 | NP_032630.2 | 18 - 19 | 1869 | 86 | ||
GenBank mRNA | AK031446.1 | - | 18 - 19 | 1870 | 86 | |
AK033824.1 | - | 18 - 19 | 1930 | 86 | ||
AK076908.1 | - | 18 - 19 | 2083 | 86 | ||
BC034092.1 | - | 18 - 19 | 1891 | 4483558 | 86 | |
BC066840.1 | - | 17 - 18 | 1778 | 86 | ||
M81591.1 | - | 17 - 18 | 1781 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |