Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039934.1 | NP_001035023.1 | 5 - 6 | 421 | 91 | |
NM_008632.2 | NP_032658.2 | 5 - 6 | 421 | 91 | ||
GenBank mRNA | AK046341.1 | - | 3 - 4 | 286 | 91 | |
AK079618.1 | - | 3 - 4 | 448 | 91 | ||
AK086484.1 | - | 3 - 4 | 285 | 91 | ||
AK141458.1 | - | 3 - 4 | 186 | 91 | ||
AK166380.1 | - | 5 - 6 | 437 | 91 | ||
BC052446.1 | - | 5 - 6 | 421 | 91 | ||
BC150945.1 | - | 5 - 6 | 420 | 9087856 | 91 | |
BU056737.1 | - | 5 - 6 | 421 | 91 | ||
M21041.1 | - | 3 - 4 | 187 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |