Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009012.2 | NP_033038.2 | 7 - 8 | 1313 | 78 | |
GenBank mRNA | AK037641.1 | - | 7 - 8 | 1314 | 78 | |
AK144155.1 | - | 7 - 8 | 1305 | 78 | ||
BC058180.1 | - | 7 - 8 | 1289 | 78 | ||
U66887.1 | - | 7 - 8 | 1291 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |