Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Relb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290457.1 | NP_001277386.1 | 4 - 5 | 782 | 142 | |
NM_009046.2 | NP_033072.2 | 5 - 6 | 791 | 142 | ||
GenBank mRNA | AK146932.1 | - | 5 - 6 | 792 | 142 | |
AK156767.1 | - | 5 - 6 | 747 | 142 | ||
BC019765.1 | - | 5 - 6 | 791 | 3584200 | 142 | |
BC034523.1 | - | 4 - 5 | 595 | 4485540 | 142 | |
BC117792.2 | - | 7 - 8 | 142 | |||
BC117793.1 | - | 5 - 6 | 727 | 40092162 | 142 | |
M83380.1 | - | 5 - 6 | 628 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |