Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Resp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009049.1 | NP_033075.1 | 4 - 5 | 464 | 72 | |
GenBank mRNA | AK012477.1 | - | 4 - 5 | 415 | 72 | |
AK019440.1 | - | 4 - 5 | 414 | 72 | ||
BC027535.1 | - | 4 - 5 | 427 | 3376899 | 72 | |
L34214.1 | - | 4 - 5 | 464 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |