Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198949.1 | NP_001185878.1 | 6 - 7 | 1647 | 78 | |
NM_009067.5 | NP_033093.2 | 6 - 7 | 1971 | 78 | ||
GenBank mRNA | AK145387.1 | - | 6 - 7 | 1511 | 78 | |
AK159404.1 | - | 6 - 7 | 1524 | 78 | ||
BC054329.1 | - | 6 - 7 | 1562 | 78 | ||
BC067073.1 | - | 5 - 6 | 1406 | 30611880 | 78 | |
BC075732.1 | - | 6 - 7 | 1596 | 6507517 | 78 | |
BC076636.1 | - | 6 - 7 | 1563 | 30658617 | 78 | |
BC138546.1 | - | 5 - 6 | 1412 | 8861566 | 78 | |
BC145320.1 | - | 4 - 5 | 1250 | 9053875 | 78 | |
BC145815.1 | - | 5 - 6 | 1412 | 40131118 | 78 | |
X80937.1 | - | 6 - 7 | 1548 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |