Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146196.1 | NP_001139668.1 | 5 - 6 | 872 | 88 | |
NM_009132.2 | NP_033158.2 | 5 - 6 | 872 | 88 | ||
GenBank mRNA | AK133484.1 | - | 5 - 6 | 828 | 88 | |
AK136080.1 | - | 3 - 4 | 414 | 88 | ||
AK167739.1 | - | 5 - 6 | 827 | 88 | ||
AK171530.1 | - | 5 - 6 | 858 | 88 | ||
AK173295.1 | - | 5 - 6 | 834 | 88 | ||
BC063328.1 | - | 5 - 6 | 827 | 30314464 | 88 | |
U04354.1 | - | 5 - 6 | 827 | 88 | ||
Y13971.1 | - | 5 - 6 | 827 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |