Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009148.3 | NP_033174.2 | 15 - 16 | 2347 | 72 | |
GenBank mRNA | AF022962.1 | - | 15 - 16 | 2373 | 72 | |
AK028250.1 | - | 13 - 14 | 2079 | 72 | ||
AK173235.1 | - | 15 - 16 | 2350 | 72 | ||
BC034644.1 | - | 15 - 16 | 2347 | 5355276 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |