Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009162.3 | NP_033188.3 | 4 - 5 | 592 | 72 | |
GenBank mRNA | AK005331.1 | - | 4 - 5 | 541 | 72 | |
AK011938.1 | - | 4 - 5 | 523 | 72 | ||
AK017481.1 | - | 4 - 5 | 645 | 72 | ||
AK019337.1 | - | 4 - 5 | 593 | 72 | ||
AK031856.1 | - | 4 - 5 | 526 | 72 | ||
AK161556.1 | - | 4 - 5 | 531 | 72 | ||
AK161579.1 | - | 4 - 5 | 533 | 72 | ||
BC029021.1 | - | 4 - 5 | 574 | 5361810 | 72 | |
X15830.1 | - | 4 - 5 | 570 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |