Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009167.3 | NP_033193.2 | 2 - 3 | 191 | 109 | |
GenBank mRNA | BC103611.1 | - | 2 - 3 | 191 | 40046304 | 109 |
BC103612.1 | - | 2 - 3 | 191 | 40046306 | 109 | |
BC105644.1 | - | 2 - 3 | 191 | 40046301 | 109 | |
BC105645.1 | - | 2 - 3 | 191 | 40046303 | 109 | |
U46854.1 | - | 2 - 3 | 191 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |