Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Soat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009230.3 | NP_033256.2 | 10 - 11 | 1171 | 103 | |
GenBank mRNA | AK052299.1 | - | 7 - 8 | 728 | 103 | |
AK142486.1 | - | 10 - 11 | 1088 | 103 | ||
AK159393.1 | - | 10 - 11 | 1101 | 103 | ||
AK159529.1 | - | 10 - 11 | 1101 | 103 | ||
AK160017.1 | - | 10 - 11 | 1101 | 103 | ||
AK194774.1 | - | 4 - 5 | 234 | 103 | ||
BC083092.1 | - | 9 - 10 | 985 | 30446741 | 103 | |
DQ903181.1 | - | 10 - 11 | 1088 | 103 | ||
S81092.1 | - | 10 - 11 | 1105 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |