Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Soat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009230.3 | NP_033256.2 | 12 - 13 | 1394 | 78 | |
GenBank mRNA | AK052299.1 | - | 9 - 10 | 951 | 78 | |
AK142486.1 | - | 12 - 13 | 1311 | 78 | ||
AK159393.1 | - | 12 - 13 | 1324 | 78 | ||
AK159529.1 | - | 12 - 13 | 1324 | 78 | ||
AK160017.1 | - | 12 - 13 | 1324 | 78 | ||
BC083092.1 | - | 11 - 12 | 1208 | 30446741 | 78 | |
DQ903181.1 | - | 12 - 13 | 1311 | 78 | ||
S81092.1 | - | 12 - 13 | 1328 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |