Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009813.2 | NP_033943.2 | 4 - 5 | 787 | 62 | |
GenBank mRNA | BC064057.1 | - | 4 - 5 | 725 | 876739 | 62 |
U93291.1 | - | 4 - 5 | 743 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |