Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkapk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010765.2 | NP_034895.1 | 11 - 12 | 1798 | 94 | |
GenBank mRNA | AF039840.1 | - | 11 - 12 | 1812 | 94 | |
AY533679.1 | - | 11 - 12 | 1106 | 94 | ||
AY533681.1 | - | 6 - 7 | 659 | 94 | ||
AY533682.1 | - | 6 - 7 | 659 | 94 | ||
BC019184.1 | - | 11 - 12 | 1220 | 5010305 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |