Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010778.3 | NP_034908.1 | 9 - 10 | 1031 | 105 | |
GenBank mRNA | AB001566.1 | - | 9 - 10 | 1008 | 105 | |
AB010919.1 | - | 9 - 10 | 1028 | 105 | ||
AB022600.1 | - | 10 - 11 | 1081 | 105 | ||
AK006632.1 | - | 9 - 10 | 1032 | 105 | ||
AK162989.1 | - | 2 - 3 | 1017 | 105 | ||
BC150660.1 | - | 9 - 10 | 1032 | 9087571 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |