Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Melk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010790.2 | NP_034920.2 | 5 - 6 | 602 | 97 | |
GenBank mRNA | AK010498.1 | - | 5 - 6 | 568 | 96 | |
AK011932.1 | - | 5 - 6 | 602 | 97 | ||
AK129076.1 | - | 5 - 6 | 555 | 97 | ||
AK145021.1 | - | 5 - 6 | 537 | 97 | ||
AK145316.1 | - | 5 - 6 | 511 | 97 | ||
AK161029.1 | - | 5 - 6 | 611 | 97 | ||
AK164138.1 | - | 5 - 6 | 569 | 97 | ||
AK167457.1 | - | 5 - 6 | 526 | 97 | ||
BC020416.1 | - | 4 - 5 | 437 | 97 | ||
BC085276.1 | - | 5 - 6 | 635 | 30443236 | 97 | |
X95351.1 | - | 5 - 6 | 540 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |