Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010832.5 | NP_034962.2 | 8 - 9 | 1172 | 88 | |
GenBank mRNA | AF117066.2 | - | 8 - 9 | 1174 | 88 | |
AK077794.1 | - | 2 - 3 | 432 | 88 | ||
AK089368.1 | - | 8 - 9 | 1255 | 88 | ||
AK146208.1 | - | 8 - 9 | 1108 | 88 | ||
AK148898.1 | - | 8 - 9 | 1150 | 87 | ||
BC010226.1 | - | 8 - 9 | 1133 | 3501126 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |