Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282000.1 | NP_001268929.1 | 12 - 13 | 1846 | 124 | |
NM_001282001.1 | NP_001268930.1 | 13 - 14 | 1945 | 124 | ||
NM_011250.4 | NP_035380.3 | 13 - 14 | 1975 | 124 | ||
GenBank mRNA | AK160027.1 | - | 13 - 14 | 1922 | 124 | |
BC020528.1 | - | 13 - 14 | 1923 | 4240592 | 124 | |
U36799.1 | - | 13 - 14 | 1845 | 124 | ||
U47333.1 | - | 12 - 13 | 1788 | 124 | ||
U50850.1 | - | 13 - 14 | 1883 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |