Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011258.2 | NP_035388.2 | 11 - 12 | 1488 | 68 | |
GenBank mRNA | AK146164.1 | - | 11 - 12 | 1483 | 68 | |
AK146314.1 | - | 11 - 12 | 1483 | 68 | ||
BC085173.1 | - | 11 - 12 | 1465 | 30549691 | 68 | |
BC085174.1 | - | 11 - 12 | 66 | |||
U01222.1 | - | 11 - 12 | 1456 | 68 | ||
U07157.1 | - | 11 - 12 | 1454 | 68 | ||
U15037.1 | - | 11 - 12 | 1489 | 68 | ||
U36441.1 | - | 11 - 12 | 1465 | 68 | ||
X72711.1 | - | 11 - 12 | 1454 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |