Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166450.2 | NP_001159922.1 | 3 - 4 | 495 | 66 | |
NM_001290703.1 | NP_001277632.1 | 5 - 6 | 684 | 66 | ||
NM_001290704.1 | NP_001277633.1 | 5 - 6 | 675 | 66 | ||
NM_001290705.1 | NP_001277634.1 | 2 - 3 | 250 | 66 | ||
NM_011398.3 | NP_035528.1 | 3 - 4 | 582 | 66 | ||
GenBank mRNA | AF076981.1 | - | 3 - 4 | 583 | 66 | |
AF155812.1 | - | 2 - 3 | 170 | 66 | ||
AF155813.1 | - | 2 - 3 | 161 | 66 | ||
AF535150.1 | - | 5 - 6 | 676 | 66 | ||
AF535151.1 | - | 5 - 6 | 685 | 66 | ||
BC048692.1 | - | 2 - 3 | 305 | 6333237 | 66 | |
CX224035.1 | - | 4 - 5 | 433 | 66 | ||
CX238926.1 | - | 3 - 4 | 347 | 66 | ||
CX244278.1 | - | 3 - 4 | 581 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |