Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stau1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109905.2 | NP_001103375.1 | 6 - 7 | 1025 | 97 | |
NM_001109906.2 | NP_001103376.1 | 6 - 7 | 1043 | 97 | ||
NM_001291160.1 | NP_001278089.1 | 6 - 7 | 1025 | 97 | ||
NM_011490.4 | NP_035620.1 | 6 - 7 | 1019 | 97 | ||
NR_111895.1 | 5 - 6 | 777 | 97 | |||
GenBank mRNA | AF061942.1 | - | 6 - 7 | 1019 | 97 | |
AF395842.1 | - | 5 - 6 | 746 | 97 | ||
AK005002.2 | - | 6 - 7 | 966 | 97 | ||
AK139532.1 | - | 6 - 7 | 972 | 97 | ||
AK145826.1 | - | 6 - 7 | 1010 | 97 | ||
AK151835.1 | - | 6 - 7 | 831 | 97 | ||
AK152269.1 | - | 6 - 7 | 837 | 97 | ||
AK152744.1 | - | 6 - 7 | 831 | 97 | ||
AK158952.1 | - | 2 - 3 | 627 | 97 | ||
AK159602.1 | - | 6 - 7 | 960 | 97 | ||
AK159781.1 | - | 6 - 7 | 995 | 97 | ||
AK160979.1 | - | 6 - 7 | 984 | 97 | ||
AK167637.1 | - | 6 - 7 | 966 | 97 | ||
AY391773.1 | - | 5 - 6 | 776 | 97 | ||
BC012959.1 | - | 6 - 7 | 989 | 4188394 | 97 | |
BC082277.1 | - | 6 - 7 | 962 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |