Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011505.2 | NP_035635.1 | 12 - 13 | 1248 | 86 | |
GenBank mRNA | AF152924.1 | - | 12 - 13 | 1238 | 86 | |
BC032881.1 | - | 12 - 13 | 1202 | 1224878 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |