Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011985.2 | NP_036115.1 | 6 - 7 | 1076 | 76 | |
GenBank mRNA | AF085742.1 | - | 6 - 7 | 1076 | 76 | |
AK146510.1 | - | 6 - 7 | 883 | 76 | ||
BC107357.1 | - | 6 - 7 | 983 | 40055939 | 76 | |
BC107358.1 | - | 6 - 7 | 941 | 40055946 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |