Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083959.1 | NP_001077428.1 | 9 - 10 | 861 | 79 | |
NM_001083960.1 | NP_001077429.1 | 9 - 10 | 822 | 79 | ||
NM_012046.2 | NP_036176.1 | 10 - 11 | 936 | 79 | ||
GenBank mRNA | AF126400.1 | - | 9 - 10 | 861 | 79 | |
AF149309.1 | - | 9 - 10 | 785 | 79 | ||
AF163053.1 | - | 9 - 10 | 822 | 79 | ||
AF163054.1 | - | 9 - 10 | 810 | 79 | ||
AF165313.1 | - | 9 - 10 | 815 | 79 | ||
AF167439.1 | - | 9 - 10 | 854 | 79 | ||
AF169386.1 | - | 10 - 11 | 933 | 79 | ||
AF173848.1 | - | 10 - 11 | 929 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |