Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Surf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013677.2 | NP_038705.2 | 6 - 7 | 802 | 82 | |
NR_073420.1 | 6 - 7 | 842 | 82 | |||
GenBank mRNA | AK002469.1 | - | 6 - 7 | 616 | 82 | |
AK154726.1 | - | 6 - 7 | 622 | 82 | ||
AK208539.1 | - | 2 - 3 | 145 | 82 | ||
BC004755.1 | - | 6 - 7 | 802 | 3584373 | 82 | |
BC052500.1 | - | 6 - 7 | 789 | 5255619 | 82 | |
CB524655.1 | - | 1 - 2 | 72 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |