Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cartpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013732.7 | NP_038760.3 | 1 - 2 | 204 | 71 | |
GenBank mRNA | BC056431.1 | - | 1 - 2 | 162 | 6817832 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |