Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160404.1 | NP_001153876.1 | 8 - 9 | 1230 | 103 | |
NM_013814.3 | NP_038842.3 | 8 - 9 | 1318 | 103 | ||
GenBank mRNA | AK083391.1 | - | 8 - 9 | 1234 | 103 | |
AK148550.1 | - | 2 - 3 | 298 | 103 | ||
BC056215.1 | - | 8 - 9 | 1235 | 30019829 | 103 | |
BC090962.1 | - | 7 - 8 | 1108 | 30626890 | 103 | |
U73820.1 | - | 6 - 7 | 983 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |