Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013908.4 | NP_038936.1 | 2 - 3 | 323 | 62 | |
GenBank mRNA | AF176520.1 | - | 2 - 3 | 284 | 62 | |
AK019715.2 | - | 2 - 3 | 324 | 62 | ||
AK154911.1 | - | 1 - 2 | 1262 | 62 | ||
BC010776.1 | - | 2 - 3 | 250 | 4211592 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |