Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015776.2 | NP_056591.1 | 4 - 5 | 182 | 64 | |
GenBank mRNA | AF180805.1 | - | 4 - 5 | 182 | 64 | |
AK003479.1 | - | 4 - 5 | 199 | 64 | ||
AK011458.1 | - | 4 - 5 | 259 | 64 | ||
BC025131.1 | - | 4 - 5 | 181 | 3982519 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |