Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016762.2 | NP_058042.2 | 2 - 3 | 396 | 68 | |
GenBank mRNA | AK035102.1 | - | 2 - 3 | 398 | 68 | |
AK081339.1 | - | 2 - 3 | 340 | 68 | ||
AK083151.1 | - | 2 - 3 | 393 | 68 | ||
AK138211.1 | - | 2 - 3 | 393 | 68 | ||
AK163818.1 | - | 2 - 3 | 355 | 68 | ||
BC005429.1 | - | 2 - 3 | 266 | 3492881 | 68 | |
BC092298.1 | - | 2 - 3 | 339 | 5063535 | 68 | |
U69262.1 | - | 2 - 3 | 395 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |