Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016762.2 | NP_058042.2 | 4 - 5 | 1079 | 74 | |
GenBank mRNA | AK083151.1 | - | 4 - 5 | 1076 | 74 | |
AK138211.1 | - | 4 - 5 | 1076 | 74 | ||
AK163818.1 | - | 4 - 5 | 1038 | 74 | ||
BC005429.1 | - | 4 - 5 | 949 | 3492881 | 74 | |
BC092298.1 | - | 4 - 5 | 1022 | 5063535 | 74 | |
U69262.1 | - | 4 - 5 | 1078 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |