Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016762.2 | NP_058042.2 | 13 - 14 | 2200 | 86 | |
GenBank mRNA | AK083151.1 | - | 13 - 14 | 2197 | 86 | |
AK138211.1 | - | 13 - 14 | 2197 | 86 | ||
AK163818.1 | - | 13 - 14 | 2159 | 86 | ||
BC005429.1 | - | 13 - 14 | 2070 | 3492881 | 86 | |
BC092298.1 | - | 13 - 14 | 2143 | 5063535 | 86 | |
U69262.1 | - | 13 - 14 | 2199 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |