Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntnap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016782.2 | NP_058062.2 | 9 - 10 | 1597 | 59 | |
GenBank mRNA | AF039833.1 | - | 9 - 10 | 1607 | 59 | |
AK154468.1 | - | 2358 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |