Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016795.3 | NP_058075.2 | 8 - 9 | 796 | 145 | |
GenBank mRNA | AB012290.1 | - | 7 - 8 | 754 | 145 | |
AJ224115.1 | - | 8 - 9 | 145 | |||
AK140761.1 | - | 206 | 145 | |||
AK151152.1 | - | 8 - 9 | 859 | 145 | ||
AK161346.1 | - | 8 - 9 | 774 | 145 | ||
BC005707.1 | - | 8 - 9 | 765 | 3157835 | 145 | |
BC050761.1 | - | 8 - 9 | 796 | 6771151 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |