Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016870.3 | NP_058566.3 | 11 - 12 | 1682 | 67 | |
NM_212441.2 | NP_997606.2 | 10 - 11 | 1577 | 67 | ||
NM_212442.2 | NP_997607.2 | 10 - 11 | 1480 | 67 | ||
GenBank mRNA | AB022340.2 | - | 10 - 11 | 1468 | 67 | |
AF068246.1 | - | 10 - 11 | 1467 | 67 | ||
AK143946.1 | - | 1480 | 67 | |||
AK165516.1 | - | 10 - 11 | 1454 | 67 | ||
AY064696.1 | - | 10 - 11 | 1553 | 67 | ||
BC015248.1 | - | 11 - 12 | 1642 | 4239288 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |