Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276315.1 | NP_001263244.1 | 11 - 12 | 2074 | 102 | |
NM_016912.2 | NP_058608.1 | 11 - 12 | 1808 | 102 | ||
NM_177270.4 | NP_796244.2 | 11 - 12 | 1808 | 102 | ||
GenBank mRNA | AB029065.1 | - | 11 - 12 | 1784 | 102 | |
AB029066.1 | - | 11 - 12 | 1772 | 102 | ||
AK030598.1 | - | 11 - 12 | 2063 | 102 | ||
AK139820.1 | - | 11 - 12 | 2017 | 102 | ||
AK144574.1 | - | 11 - 12 | 1809 | 102 | ||
AK211201.1 | - | 1 - 2 | 61 | 102 | ||
BC117894.1 | - | 11 - 12 | 1717 | 40096852 | 102 | |
BC117895.1 | - | 11 - 12 | 1717 | 40096853 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |