Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168568.1 | NP_001162040.1 | 7 - 8 | 943 | 93 | |
NM_001168569.1 | NP_001162041.1 | 7 - 8 | 1000 | 93 | ||
NM_001168571.1 | NP_001162043.1 | 7 - 8 | 1075 | 93 | ||
NM_018737.5 | NP_061207.1 | 7 - 8 | 1075 | 93 | ||
NR_033143.1 | 7 - 8 | 965 | 93 | |||
GenBank mRNA | AK032018.1 | - | 7 - 8 | 802 | 93 | |
AK052854.1 | - | 7 - 8 | 991 | 93 | ||
AK084595.1 | - | 7 - 8 | 872 | 93 | ||
AK150966.1 | - | 7 - 8 | 942 | 93 | ||
AK151597.1 | - | 7 - 8 | 942 | 93 | ||
AK161564.1 | - | 7 - 8 | 913 | 93 | ||
AK164206.1 | - | 7 - 8 | 902 | 93 | ||
BC003257.1 | - | 7 - 8 | 783 | 3500778 | 93 | |
U49385.1 | - | 7 - 8 | 965 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |