Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300008513.mm.1 |
NM_001276676.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008513.mm.1 |
NM_001276677.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008513.mm.1 |
NM_001276679.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008513.mm.1 |
NM_018800.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008513.mm.1 |
TC0300001055.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008521.mm.1 |
NM_001276676.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008521.mm.1 |
NM_001276677.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008521.mm.1 |
NM_001276679.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008521.mm.1 |
NM_001276680.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008521.mm.1 |
NM_001276681.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008521.mm.1 |
NM_018800.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008521.mm.1 |
TC0300001055.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008512.mm.1 |
NM_001276676.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008512.mm.1 |
NM_001276677.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008512.mm.1 |
NM_001276679.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008512.mm.1 |
NM_018800.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008512.mm.1 |
TC0300001055.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008520.mm.1 |
NM_001276680.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008520.mm.1 |
NM_018800.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008520.mm.1 |
TC0300001055.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008523.mm.1 |
NM_001276676.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008523.mm.1 |
NM_001276677.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008523.mm.1 |
NM_001276679.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008523.mm.1 |
NM_001276680.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008523.mm.1 |
NM_001276681.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008523.mm.1 |
NM_018800.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008523.mm.1 |
TC0300001055.mm.1 |
MTA TC |