Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019521.2 | NP_062394.2 | 1 - 2 | 273 | 66 | |
GenBank mRNA | AK086187.1 | - | 1 - 2 | 267 | 66 | |
BC005444.1 | - | 1 - 2 | 228 | 3592398 | 66 | |
CJ062651.1 | - | 1 - 2 | 274 | 66 | ||
X59846.1 | - | 1 - 2 | 244 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |