Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maged1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019791.2 | NP_062765.1 | 8 - 9 | 1942 | 85 | |
GenBank mRNA | AB029448.1 | - | 8 - 9 | 1942 | 85 | |
AF319975.1 | - | 7 - 8 | 1712 | 85 | ||
AK013231.1 | - | 1 - 2 | 161 | 85 | ||
AK017275.1 | - | 8 - 9 | 1860 | 85 | ||
AK049660.1 | - | 8 - 9 | 1893 | 85 | ||
AK087937.1 | - | 8 - 9 | 1892 | 85 | ||
AK132248.1 | - | 8 - 9 | 1881 | 85 | ||
AK135452.1 | - | 8 - 9 | 1892 | 85 | ||
AK145872.1 | - | 8 - 9 | 1862 | 85 | ||
AK146867.1 | - | 8 - 9 | 1892 | 85 | ||
AK146908.1 | - | 8 - 9 | 1892 | 85 | ||
AK150876.1 | - | 5 - 6 | 881 | 85 | ||
AK220150.1 | - | 9 - 10 | 1799 | 85 | ||
BC016438.1 | - | 7 - 8 | 2076 | 4480947 | 85 | |
BC031461.1 | - | 8 - 9 | 1882 | 4018875 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |