Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159589.1 | NP_001153061.1 | 5 - 6 | 1093 | 67 | |
NM_019812.2 | NP_062786.1 | 6 - 7 | 1207 | 67 | ||
GenBank mRNA | AF214646.1 | - | 6 - 7 | 1192 | 67 | |
AK144203.1 | - | 5 - 6 | 1096 | 67 | ||
AY377984.1 | - | 6 - 7 | 1192 | 67 | ||
BC152314.1 | - | 6 - 7 | 687 | 40111573 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |