Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mast1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019945.2 | NP_064329.2 | 19 - 20 | 2397 | 91 | |
GenBank mRNA | AF077818.1 | - | 19 - 20 | 2380 | 91 | |
AK122411.1 | - | 9 - 10 | 1532 | 91 | ||
BC039230.1 | - | 8 - 9 | 1382 | 91 | ||
BC054524.1 | - | 19 - 20 | 2397 | 6400705 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |