Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600011320.mm.1 |
TC0600001403.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011320.mm.1 |
NM_001190320.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011320.mm.1 |
NM_001190320.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011320.mm.1 |
NM_001190321.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011320.mm.1 |
NM_001190321.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011320.mm.1 |
NM_020001.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011320.mm.1 |
NM_020001.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011326.mm.1 |
TC0600001403.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011326.mm.1 |
NM_001190320.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011326.mm.1 |
NM_001190320.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011326.mm.1 |
NM_020001.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011326.mm.1 |
NM_020001.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011322.mm.1 |
TC0600001403.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011322.mm.1 |
NM_001190321.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011322.mm.1 |
NM_001190321.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011322.mm.1 |
NM_020001.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011322.mm.1 |
NM_020001.2 |
Refseq |